Learn More
Advanced Chemblocks Inc JQ 1
Supplier: Advanced Chemblocks Inc L14965100MG
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Advanced ChemBlocks Inc, jq 1, Cat ID: L14965, CAS: 1268524-70-4, MDL: MFCD22683748, FW: 457, Formula: C23H25CLN4O2S, IUPAC Name: rac-tert-butyl (r)-2-(4-(4-chlorophenyl)-2,3,9-trimethyl-6h-thieno[3,2-f][1,2,4]triazolo[4,3-a][1,4]diazepin-6-yl)acetate