Learn More
Kurin Jet (Blood Culture Collection Device)
Jet is a blood culture (BC) collection device which significantly lowers BC contamination rates. Jet automatically sidelines the initial 0.15 mL of blood, which may contain contaminants from the patient’s own skin, ensuring that the best possible blood specimen reaches the collection bottles.
Supplier: Kurin MJPIV12
Description
- Lowers blood culture contamination rate
- Helps achieve <1% contamination rate
- Minimal blood volume waste
- Closed System - All-In-One Device
- Fits BD, bioMerieux & ThermoFisher Blood Culture Bottles
Specifications
Jet for IV Connection for BM Blood Culture Bottles (case) | |
Blood Culture Collection | |
200 per Cs. | |
IV Connection, Kurin Jet, Vacutainer Shield to fit BM BC Bottles |