Learn More
Glass Expansion IsoMist XS Kit for Thermo PRO
Supplier:  Glass Expansion KT1166XS
Encompass_Preferred
IsoMist XS Kit Thermo PRO. Compatible with Thermo PRO ICP-OES. This IsoMist XS is programmable and temperature-controlled, with a glass cyclonic spray chamber now featuring an improved thermodynamic design that provides an extended temperature range of -25°C to +80°C. The temperature is settable in 1°C increments and can maintain accuracy of + or – 0.1°C. Proven performance results are, improved long-term stability and precision, no dilution for volatile organic solvents, reduced oxide interferences (ICP-MS), enhanced sensitivity and accuracy, lower detection limits, faster washout, and reduced hazardous waste/disposal fees.Replacement parts include 21-809-3186, NC1943061, and 501907304.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-2970