Learn More
Regis Technologies Inc Hexyltriethylammonium Phosphate Buffer, 100ml
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Regis Technologies Inc 1404036200
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The Q-Series is comprised of quaternary alkyltriethylamines that can be used for the resolution of negatively charged species. The Q-Series Ion Pair Concentrates are available as premixed 0.5 M solutions of varying alkyl chain lengths. When 10 mL of concentrate is diluted to 1 L with HPLC-grade water, a 0.005 M buffered solution is formed.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.