Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Iodine, crystalline, 99.5%
Specifications
Melting Point | 183-186° subl. |
---|---|
CAS | 7553-56-2 |
Physical Form | Solid |
Molecular Formula | I{2} |
MDL Number | MFCD00011355 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AA1424822
|
Thermo Scientific™
1424822 |
100 g |
Each for $77.80
Order in Bulk
Request bulk or custom formats |
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
AA1424828
|
Thermo Scientific™
1424828 |
200 g |
Each for $138.00
Order in Bulk
Request bulk or custom formats |
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
AA14248A1
|
Thermo Scientific™
14248A1 |
1 kg |
Each
for $546.65
Save 8% Reg :
Order in Bulk
Request bulk or custom formats |
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
Specifications
183-186° subl. | |
Solid | |
MFCD00011355 | |
253.81 | |
4.930 |
7553-56-2 | |
I{2} | |
UN3495 | |
99.5% |
Safety and Handling
ShelfLife : 5 Years
EINECSNumber : 231-442-4
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title