Learn More
Chemglass Life Sciences Internal Bushing, #25, PTFE, 1/8" NPT, #212 Viton O-Ring
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Chemglass Life Sciences CG36460
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Internal Bushing, #25, PTFE, 1/8" NPT, #212 Viton O-Ring. New front seal PTFE bushing with female NPT for use with CG-360 internal threads. Supplied complete with Viton o-ring of the listed size. See CG-305 for Viton and CG-309 for Perfluoro replacement o-rings.