Learn More
Revvity Health Sciences Inc Insulin Mouse Serum Assay kit, 1000 Tests
Supplier: Revvity Health Sciences Inc 62IN3PEB
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
For years, researchers have sought the ideal insulin assay that would combine absolute specificity with sensitivity, accuracy, low sample consumption, and ease-of-use. The search has come to an end! This serum insulin assay offers superior specificity and sensitivity for a reliable quantification of insulin in serum and plasma samples. Built on a truly homogeneous protocol, the assay does not require any washing or complex extraction steps, leading to significant time savings over ELISA methods.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.