Learn More
Restek™ Instrument Grade Tubing
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Used for providing carrier, fuel, make-up, or auxiliary gases to laboratory instruments.
Supplier: Restek™ 21514
Description
Note: Quantity of Each is the price per foot. Minimum order quantity is 100 ft.Order Info
Maximum continuous lengths are: 2000 ft. (1/16 in. OD), 1500 ft. (1/8 in. OD), and 750 ft. (1/4 in. OD). The availability of long lengths is subject to inventory constraints. Lead times may vary depending on the continuous length needed. Please inquire before ordering.
Specifications
Welded/Drawn Tubing | |
For Providing Carrier, Fuel, Make-up or Auxiliary Gases to Laboratory Instruments | |
0.125 in. | |
100 to 500 ft. | |
Stainless Steel (304) |
CGA | |
0.85 in. | |
0.317 cm | |
30.48 to 152.4 m |