Learn More
METTLER TOLEDO CO2 Sensor InPro5000i/12/220
Supplier: METTLER TOLEDO 30019005
![Encompass](/content/dam/fishersci/glyphs/encompass.png)
The InPro 5000i 12/220 is an on-line carbon dioxide sensor with ISM that allows for accurate measurement and control of dissolved CO2 in biopharmaceutical applications. It is based on the widely accepted Severinghaus principle of CO2 measurement.
Description
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)