Learn More
Ace Glass, Inc. Impinger, 25mL, complete with 12/5 socket inlet, 12/5 ball outlet stopper, open tube end nozzle
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Ace Glass, Inc. 753137
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
25mL midget impinger used for sampling small air volumes at low velocities. Includes 12/5 socket inlet and 12/5 ball outlet stopper with an open tube end nozzle.