Learn More
Cole-Parmer George Ulanet 335-115 Heat-O-Matic Immersion Heater, loop, 3-1/8" dia. Heated Area, 500 watts, 115 VAC
Supplier:  Cole-Parmer 0304620
Encompass_Preferred
Heaters with Dial Temperature Controller have a sensitivity of ±1°>F when liquid is stirred. Top-mounted pilot light indicates that heater is on. Choose from four styles "A" loop, "B" short rod, "C" flat, and "D" long rod which is the same as style "B" only longer. Heaters feature a plastic grip handle; and a 6-ft, three-wire cord, 115 VAC models have U.S. standard plug, 230 VAC models do not have a plug.
Catalog No. NC9116074
Explore available promotions