Learn More
TARGETMOL CHEMICALS INC IMIDAZOLE KETONE ERASTIN 5MG
Supplier: TARGETMOL CHEMICALS INC T5523
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Imidazole ketone erastin (IKE) (IKE) is an inducer of ferroptosis. It has inhibition of the system Xc- cystine/glutamate transporter. purity: 99%