Learn More
Electron Microscopy Sciences Ilford Ilfospeed RC Deluxe Paper, Glossy-Grade 3
Supplier: Electron Microscopy Sciences 8303325
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
This is a direct replacement for Kodak Kodabrome II RC B&W Papers. Premium Quality Graded Paper on a Resin Coated Base. MULTIGRADE RC COOLTONE Paper has the standard weight of (190g/m 2) with a resin coated base. It is suitable for a wide range of applications including commercial, press, industrial, advertising, and display work.
Grade: Glossy-Grade 3
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.