Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ICP Instrument Check Standard, Solution II, Spectrum™ Chemical
![SDP](/content/dam/fishersci/glyphs/sdp.png)
$279.00 - $735.00
Specifications
Packaging | Poly Bottle |
---|---|
Type | ICP Instrument Check Standard |
Grade | Chemical Solution |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
18-611-005
![]() |
Spectrum Chemical Manufacturing Corporation
PM430100ML |
100 mL | Poly Bottle |
Each for $279.00
|
|
||||
18-611-006
![]() |
Spectrum Chemical Manufacturing Corporation
PM430500ML |
500 mL | Poly Bottle |
Each for $735.00
|
|
||||
Specifications
Poly Bottle | |
ICP Instrument Check Standard | |
Chemical Solution |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title