Learn More
TARGETMOL CHEMICALS INC IBUTILIDE FUMARATE 100MG
Supplier: TARGETMOL CHEMICALS INC T6541
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Also available in 10 mg 25 mg 50 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ibutilide Fumarate (U70226E) is the fumarate salt form of ibutilide a class III antiarrhythmic agent. Ibutilide exerts its effect by activating a slow inward predominately sodium current rather than by blocking outward potassium currents. This results in prolongation of atrial and ventricular action potential duration and refractory periods. Ibutilide slightly decreases the sinus rate and atrioventricular (AV) conduction and produces a dose-related prolongation of the QT interval. purity: 99%
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.