Learn More
TARGETMOL CHEMICALS INC HYPOCRELLIN A 5MG
Supplier: TARGETMOL CHEMICALS INC T5771
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Hypocrellin A is PKC inhibitor has light-induced antitumorantifungal and antiviral activities it also exerts immunomodulatory effects on MHC-restricted presentation of antigen. purity: 98%