Learn More
Novus Biologicals™ Hydroxyproline Assay Kit (Colorimetric)
SDP
This kit uses an acidic developer solution to accurately measure hydroxyproline in tissue and protein/peptidase hydrolysates without the use of toxic perchlorates.
Supplier:  Novus Biologicals™ NBP259747
Catalog No. NBP259747
Explore available promotions
Add to Cart
Description
Description
Featuring a simple and quick protocol, this hydroxyproline assay yields a brightly-colored chromophore with an absorbance peak at 560 nm. The assay can detect a minimum of 0.1 μg hydroxyproline per well in a 96-well format and generates colorimetric results that are virtually identical to the classical perchloric acid-based method.
Kit Contents:
  • Oxidation Buffer
  • DMAB Concentrate (in DMSO)
  • Developer Solution
  • Microplate Sealing Film
  • Hydroxyproline Standard (1 mg/mL)
  • Chloramine T Concentrate
Specifications
Specifications
100 Assays
-20°C
Hydroxyproline
Product Suggestions
Product Suggestions