Learn More
STEMCELL Technologies Human Peripheral Blood Plasma, Frozen, Size: 10 mL
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: STEMCELL Technologies 70039.1
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Primary human plasma has been isolated from peripheral blood (PB) using centrifugation. PB was collected using one of the following anticoagulants: acid-citrate-dextrose (ACD), acid-citrate-dextrose solution A (ACDA), citrate-phosphate-dextrose (CPD), citrate-phosphate-double-dextrose (CP2D), or citrate-phosphate-dextrose-adenine (CPDA).Plasma was obtained using Institutional Review Board (IRB)-approved consent forms and protocols.Certain products are only available in select territories. Please contact your local Sales representative or Product & Scientific Support at techsupport@stemcell.com for further information.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.