Learn More
Sino Biological Human MMP12 / MMP-12 / HME Protein (catalytic domain) 1mg
Supplier: Sino Biological 10266HNAE1
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
A DNA sequence encoding human MMP12 (NP_002417.2) (Gly106-Asn268) was expressed with a N-terminal Met.
This item is not returnable.
View return policy