Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Human IL-1 beta/IL-1F2 Quantikine ELISA Kit
Human IL-1 beta/IL-1F2 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Supplier: R&D Systems™ SLB50
This item is not returnable.
View return policy
Description
The Human IL-1 beta/IL-1F2 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human IL-1 beta/IL-1F2 in cell culture supernates (200 ul), serum (200 ul), edta plasma (200 ul), heparin plasma (200 ul), citrate plasma (200 ul).Specifications
3.90 - 250pg/mL | |
HRP | |
Solid Phase Sandwich ELISA | |
Natural and Recombinant Human IL-1 beta | |
Human | |
3553 | |
Cell Culture Supernates (200uL),Serum (200uL),EDTA Plasma (200uL),Heparin Plasma (200uL),Citrate Plasma (200uL) | |
catabolin, IL-1, IL1 beta, IL-1 beta, IL-1 b, IL1B, IL1-BETA, IL-1F2, IL1F2IL-1 beta, interleukin 1, beta, interleukin-1 beta, preinterleukin 1 beta, pro-interleukin-1-beta |
1pg/mL | |
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules | |
Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum | |
Sufficient materials to run ELISAs on at least six 96-well plates | |
ELISA | |
6 Plates | |
Store the unopened product at 2° to 8°C. Do not use past expiration date. |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction