Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IgG2 Isotype Control, Novus Biologicals™
Human Polyclonal Antibody
Supplier: Novus Biologicals DDXCH02P100
Description
IgG2 Polyclonal specifically detects IgG2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Cell Surface), Flow (Intracellular), Immunofluorescence.Specifications
IgG2 | |
Polyclonal | |
Unconjugated | |
Tris-NaCl, pH 8. with No Preservative | |
IGHG2 | |
Ion exchange chromatography | |
RUO | |
Isotype Control (Primary) | |
Human | |
IgG2 |
Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Flow Cytometry, Immunofluorescence | |
0.5 mg/mL | |
Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Flow (Cell Surface) 1:10 - 1:1000, Flow (Intracellular) 1:10 - 1:1000, Immunofluorescence 1:10 - 1:500 | |
Constant region of heavy chain of IgG2, immunoglobulin gamma 2 (Gm marker), immunoglobulin heavy constant gamma 2 (Gm marker) | |
Human | |
0.1 mg | |
Immunology | |
3501 | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction