Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IgG1 Isotype Control, Novus Biologicals™
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Human Polyclonal Antibody has been used in 1 publication
Supplier: Novus Biologicals DDXCH01P100
Description
IgG1 Polyclonal specifically detects IgG1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Cell Surface), Flow (Intracellular), Immunofluorescence.Specifications
IgG1 | |
Polyclonal | |
Unconjugated | |
Tris-NaCl, pH 8. with No Preservative | |
IGHG1 | |
Ion exchange chromatography | |
RUO | |
Isotype Control (Primary) | |
Store at -20C. Avoid freeze-thaw cycles. |
Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Flow Cytometry, Immunofluorescence | |
0.5 mg/mL | |
Immunohistochemistry-Paraffin 1:10 - 1:500, Immunohistochemistry-Frozen 1:10 - 1:500, Flow (Cell Surface) 1:10 - 1:1000, Flow (Intracellular) 1:10 - 1:1000, Immunofluorescence 1:10 - 1:500 | |
IgG1, Igh-4, RGD1359539, VH7183 | |
Human | |
0.1 mg | |
Immunology | |
Human | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction