Learn More
Rockland Immunochemicals Human Blood in Alsevers 50 mL
SDP
Supplier:  Rockland Immunochemicals R3140050
Encompass_Preferred
Human whole blood is aseptically collected and processed upon receipt of your order. These cell suspensions are useful for the titration of complement adsorption procedures testing for agglutinins/HA assays and for the preparation of stroma as particulate reagents. Human red blood cells are perishable and are collected and processed upon receipt of your order.
Catalog No. 50-194-3986
Explore available promotions