Learn More
Foxx Life Sciences EZWaste® HPLC VersaCap® 53m (53B) Solvent Waste System, Cap-Only W/ Exhaust Filter, Six (6x) Ports for OD Tubing 3.2 mm (1/8"") or 1.6 mm (1/16""), 1/EA
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Foxx Life Sciences 3300212FLS
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Foxx Life Sciences EZWaste® HPLC VersaCap® 53mm Solvent Waste Cap Assembly is designed to be an affordable solution for HPLC liquid waste disposal. Each system is manufactured to safely collect waste from multiple HPLC machines. This system features an activated carbon filter which traps volatile organic compounds from being released into the air. This prevents chemicals from being inhaled by lab workers and provides safer mechanism of disposal for vapor wastes. The EZwaste® Solvent Waste system also features a closed loop design which protects against leaks and spills, improving overall lab safety. Each EZwaste® Assembly includes a VersaCap® 53mm with Manifold, fittings 6 x 3.2 mm (1/8") OD Tubing, fittings 6 x 1.6 mm (1/16") OD Tubing, 6 x Plugs suitable for blocking of unused ports & 1 carbon exhaust filter. * Depending on application, carbon exhaust filter life cycle may vary.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.