Learn More
Sterlitech Corporation HP4750 Membrane Area Reducer, 25mm, includes Porous Support Disc and O-ring
SDP
Supplier:  Sterlitech Corporation 1140144
Encompass_Preferred
HP4750 Membrane Area Reducer, 25mm, includes Porous Support Disc and O-ring.
Membrane active area using this reducer is 0.35 in2 (2.24 cm2).
Catalog No. 50-195-2341
Explore available promotions