Learn More
Augustine Temperature Mgmt Hot Dog HOTDOG EXAM TABLE COVERS 45/CS
SDP
Supplier:  Augustine Temperature Mgmt Hot Dog V501
Encompass
HotDog Exam Table Covers feature a proprietary adhesive foam material with an adhesive backing that sticks to the exam table but leaves no residue when removed. The Exam Table Covers can be used on most surfaces, including Formica and stainless steel. They provide a non-slip surface for pets along with protection from the cold table surface. Because of the reasonable cost per sheet, it can be replaced at the end of the day or week as needed. The 20″x40″ sheets are waterproof and can be cleaned using your current disinfectant. They fit most exam tables and can be easily trimmed for use on scales.
Catalog No. NC2162660