Learn More
Electron Microscopy Sciences Combination Holder 45 ° Pin Mount and 90° Profile, Brass
Supplier: Electron Microscopy Sciences 7595007B
For Hitachi S-800, S-4000, S-4100, S-4200, S-4300, S-4500, S-4700 and S-3600N SEMs. For cross sections up to 6.35 mm ( 1/4"). Available in aluminum or brass with stainless steel allen set screws. Includes allen wrench.