Learn More
TARGETMOL CHEMICALS INC HJ-PI01 5MG
Supplier: TARGETMOL CHEMICALS INC T9583
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. HJ-PI01 (HJ-PI01) is a Pim-2 inhibitor. HJ-PI01 (HJ-PI01) induces apoptosis and autophagic cell death in triple-negative human breast cancer. purity: 99%