Learn More
Electron Microscopy Sciences EMS Glass Knife Strip Histology Grade 10 mm thick, 25 x 400 mm
Supplier: Electron Microscopy Sciences 71018
Specially selected glass strips for making glass knives. Its hardness produces a durable and sharp edge suitable for thick, semi thin, or ultrathin sections.
Thickness: 10 mm
Size: 25 x 400 mm