Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva His MultiTrap™ 96-well Filter Plates
Prepacked multiwell filter plates for high reproducibility in expression screening and small-scale purification of histidine-tagged proteins
$1345.00 - $1537.00
Specifications
Description | His MultiTrap FF and His MultiTrap HP are prepacked disposable 96-well plates for reproducible high throughput parallel purification of histidine-tagged recombinant proteins by Immobilized Metal ion Affinity Chromatography (IMAC) |
---|---|
Material | Polypropylene, Polyethylene |
pH Range | 3 to 12 |
Content And Storage | 4°C to 30°C |
Quantity | 4 x 96-well Filter Plates |
Catalog Number | Mfr. No. | Type | Particle Size | Support Type | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Particle Size | Support Type | Price | Quantity & Availability | |||
45-001-424
|
Cytiva
28400989 |
96-well Filter Plate | 34 μm | 6% Highly Cross-linked Agarose |
Pack of 4 for $1,537.00
|
|
|||
45-001-425
|
Cytiva
28400990 |
96-well Filter Plate | 45 to 165 μm | 6% Cross-linked Agarose |
Pack of 4 for $1,345.00
|
|
|||
Description
- No filtration needed: load unclarified sample directly, increasing reproducibility and saving time
- Compatible with robotic systems or manual protocols with centrifuge or vacuum
- Easy scale-up
- Long-term pH range 3 to 12
- Protein binding capacity up to 800μg histidine-tagged protein/well with His MultiTrap FF
- Protein binding capacity up to 1mg histidine-tagged protein/well with His MultiTrap HP
Specifications
His MultiTrap FF and His MultiTrap HP are prepacked disposable 96-well plates for reproducible high throughput parallel purification of histidine-tagged recombinant proteins by Immobilized Metal ion Affinity Chromatography (IMAC) | |
3 to 12 | |
4 x 96-well Filter Plates |
Polypropylene, Polyethylene | |
4°C to 30°C | |
96-well Filter Plate |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title