Learn More
Ampco Safety Tools Hex Key Wrench (Metric)
![SDP](/content/dam/fishersci/glyphs/sdp.png)
AMPCO™Hex Key Wrench performs in high-risk and demanding environments.
$8.11 - $1040.00
Specifications
Product Type | Hex Key |
---|---|
Certifications/Compliance | Factory Mutual (FM) Approved |
Alert:
AMPCO and other copper-based tools should not be used in direct contact with acetylene, ethylene oxide and some explosives such as amatol, DBX and lead azide.
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
19-174-750
![]() |
Ampco Safety Tools
WH1.5MM |
1.5 mm |
Each for $8.11
|
|
|||||
19-174-753
![]() |
Ampco Safety Tools
WH10MM |
10 mm |
Each for $72.70
|
|
|||||
19-174-754
![]() |
Ampco Safety Tools
WH11MM |
11 mm |
Each for $84.70
|
|
|||||
19-174-755
![]() |
Ampco Safety Tools
WH12MM |
12 mm |
Each for $105.10
|
|
|||||
19-174-756
![]() |
Ampco Safety Tools
WH13MM |
13 mm |
Each for $127.50
|
|
|||||
19-174-757
![]() |
Ampco Safety Tools
WH14MM |
14 mm |
Each for $126.50
|
|
|||||
19-174-758
![]() |
Ampco Safety Tools
WH17MM |
17 mm |
Each for $215.50
|
|
|||||
19-174-759
![]() |
Ampco Safety Tools
WH19MM |
19 mm |
Each for $277.50
|
|
|||||
19-174-760
![]() |
Ampco Safety Tools
WH2.5MM |
2.5 mm |
Each for $18.36
|
|
|||||
19-174-761
![]() |
Ampco Safety Tools
WH22MM |
22 mm |
Each for $310.50
|
|
|||||
19-174-762
![]() |
Ampco Safety Tools
WH24MM |
24 mm |
Each for $315.50
|
|
|||||
19-174-763
![]() |
Ampco Safety Tools
WH27MM |
27 mm |
Each for $535.50
|
|
|||||
19-174-764
![]() |
Ampco Safety Tools
WH2MM |
2 mm |
Each for $19.89
|
|
|||||
19-174-767
![]() |
Ampco Safety Tools
WH30MM |
30 mm |
Each for $627.50
|
|
|||||
19-174-768
![]() |
Ampco Safety Tools
WH32MM |
32 mm |
Each for $806.00
|
|
|||||
19-174-769
![]() |
Ampco Safety Tools
WH36MM |
36 mm |
Each for $1,040.00
|
|
|||||
19-174-770
![]() |
Ampco Safety Tools
WH3MM |
3 mm |
Each for $19.64
|
|
|||||
19-174-771
![]() |
Ampco Safety Tools
WH4MM |
4 mm |
Each for $20.15
|
|
|||||
19-174-774
![]() |
Ampco Safety Tools
WH5MM |
5 mm |
Each for $25.00
|
|
|||||
19-174-775
![]() |
Ampco Safety Tools
WH6MM |
6 mm |
Each for $36.25
|
|
|||||
19-174-777
![]() |
Ampco Safety Tools
WH7MM |
7 mm |
Each for $44.40
|
|
|||||
19-174-778
![]() |
Ampco Safety Tools
WH8MM |
8 mm |
Each for $54.60
|
|
|||||
19-174-779
![]() |
Ampco Safety Tools
WH9MM |
9 mm |
Each for $57.90
|
|
|||||
Description
- Made of AMPCO invented, Factory Mutual approved alloy
- Designed for use near flammable liquids, vapors, explosive dust, sensitive electronics, magnetic equipment, batteries and in laboratories.
- Non-Sparking
- Non-Magnetic
- Corrosion Resistant
Warranty and Services
Limited Lifetime Warranty
Specifications
Hex Key | |
Factory Mutual (FM) Approved |
Common applications include maintenance repair operations (MRO) around flammable liquids, dusts and vaports. Non-magnetic environments for MRI maintenanance and other precision magnetic and radar equipment; Laboratories, food and beverage processing, pharmaceuticals and chemicals handling and processing where corrosion resistant hand tools are needed.
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.