Learn More
Cole-Parmer George Ulanet 492-2 Heat-O-Matic Immersion Heater with Thermostatic Controller, 1500 watts, 7" dia
Supplier: Cole-Parmer 0305220
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Heaters with Thermostatic Temperature Controller have a sensitivity of ±2°>F(±1°>C) in an agitated bath. Heaters feature a coiled heating element to concentrate heat at bottom of vessel. A 6-ft, three-wire cord with plug is included. Voltage transformer listed below is required for 230 VAC operation.