Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
International Enviroguard™ Heavyweight Polypropylene Coveralls with Hood and Boot
Click to view available options
Size:
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Description
- With attached hood and boots for full body protection
- Low-cost protection against dirt and grime
- Made with heavyweight fabric for enhanced safety
- Possible applications include food processing, construction, pharmaceuticals, environmental cleanup, and more
Specifications
Product Type | Light-duty Coveralls |
Size | 2X-Large |
Gender | Unisex |
Material | Polypropylene (Heavy-duty) |
Color | White |
Disposable | Single-use |
Waist Style | Standard |
Ankle Style | Attached Boots |
Cuff Style | Elastic |
Neck Style | Hooded |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction