Learn More
Chemglass Life Sciences HEATER BLOCK ADAPTER
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Chemglass Life Sciences OP670001
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Heater Block OptiChem adapter is designed to fit all hot plate stirrers with a 135mm diameter top plate.