Learn More
Chemglass Life Sciences Heat Gun, Heavy-Duty, Multiple Use, 426°C Temp, 120V, 60Hz, 12A, 1440W
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Chemglass Life Sciences CG9001D02
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Professional, heavy-duty, multiple use heat gun has a sturdy die cast housing and adjustable non-slip stand. Used to bend plastics, cure adhesives, dry parts, heat parts, shrink tubing and packaging, strip paint and more. Heat gun accommodates many industrial and commercial heating applications.