Learn More
Glass Expansion High Efficiency Sample Introduction System (HE-SIS) for PerkinElmer NexION 300/350 ICP-MS.
Supplier: Glass Expansion KT1204
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
HE-SIS for PerkinElmer ICP-MS. High Efficiency Sample Introduction System (HE-SIS) for PerkinElmer NexION 300/350 ICP-MS. Originally designed as a highly efficient single-cell sample introduction system, Glass Expansion’s HE-SIS has been redesigned to provide superior performance across a wide variety of applications, including single-cell, single particle, nanoparticle, and low-volume sample studies, with up to 95% transport efficiency. Conventional ICP-MS spray chambers have low transport efficiencies (typically <5% TE) that filter out larger droplets (> 5um) from the nebulizer aerosol. The Glass Expansion HE-SIS features three critical components to overcome this significant sample transport deficiency: A High Efficiency MicroMist Nebulizer, Patent-pending MicroJet Gas Adaptor, & a High Efficiency Spray Chamber. This component combination improves the transport efficiency for superior performance.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.