Learn More
Cole-Parmer HI-200D-HC-T-120 Hybridization Incubator with Tubes, 100°C; 120 VAC
Supplier:  Cole-Parmer 3945501
Encompass_Preferred
With excellent stability of ±0.1°C within the tubes, these hybridization incubators are great for nucleic hybridization applications such as Northern, Southern, and Western blots. Each incubator features two shakers—rotisserie shaker with rotational speed from 0 to 20 rpm and platform shaker with rocking speed up to 60 oscillation/minute. A removable drip tray provides for easy cleaning. Adjustable feet make leveling on uneven surfaces quick and easy.
Catalog No. NC2064631
Explore available promotions