Learn More
Haztech Systems, Inc HAZCAT 2.0 Pro Kit
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Supplier: Haztech Systems, Inc KT7003
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
![Request Bulk Order](/content/dam/fishersci/glyphs/15395_Quote_Bulk_EN.jpg)
Industrial Chemicals/WMD/Narcotics Testing System. Hazcat's 2.0 KT7003 Pro Kit quickly & easily Identifies mixed wastes, Household products, Commercial formulations, Heavy metals, Chemical and Biological warfare agents and their precursors, Narcotics and their precursors, Tics & Tims and Industrial Poisons