Learn More
Electron Microscopy Sciences Harris Uni-Core™ Sampling Tool 2.0
Supplier: Electron Microscopy Sciences 6903620
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The Harris Uni-Core is a disposable multi-purpose sampling tool for soft substrates. Made from polypropylene, including a hollow body with built-in tool steel razor sharp cutting tip, with a plunger for the expelling of the punched sample. Uni-Core comes individually wrapped and sterilized by ethylene oxide. Uni-Core can also be reusable by rinsing the tip with ethanol after each use and autoclaving for 20 minutes at 250 degree F/15 psi.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.