Learn More
Perkin Elmer US LLC Nichrome Hangdown Wire Kit - 0.01 OD, Qty 5
Supplier: Perkin Elmer US LLC N5320131
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Hangdown wires suspend down from the TGA 8000 Furnace Balance Mechanism and catch a sample pan's hook and lift the pan from the Autosampler and into the furnace. The hangdown wires come in a variety of materials and kit sizes.