Learn More
MilliporeSigma™ Supelco™ Hamilton™ Needles for RN Syringes, Blunt Tip, Stainless Steel
Hamilton™ needles for RN syringes are bevel tip needles, designed for optimum septum penetration and to prevent septum coring.
Supplier:  MilliporeSigma™ Supelco™ 19142U
 View more versions of this product
Catalog No. 11-101-1704
Item Discontinued Due to product restrictions, please to purchase or view availability for this product.This item has been discontinued by the supplier.
View Alternate Product
Explore available promotions
Description
Description
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Stainless Steel (Tip)
2 in.
6/Pk.
51 mm
22s G
Blunt Tip
250 μL to 10 mL
Hamilton RN Syringes
Product Suggestions
Product Suggestions
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hamilton is a registered trademark of Hamilton Co.