Learn More
Educational Innovations GREEN FLUORSC MAGIC SAND
Supplier: Educational Innovations MS3GRN
Magic Sand is regular sand which has been dyed and coated with a hydrophobic substance - a substance which repels water. The coating on the outside of the magic sand repels water and keeps the sand dry!