Learn More
Electron Microscopy Sciences Graphene-Enhanced Lacey Carbon on 200 mesh Nickel Grid
Supplier: Electron Microscopy Sciences GF1202
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Graphene is the ideal support film for electron microscopy. It provides an invisible, crystalline background that enables the unrivaled TEM characterization of organic and inorganic nanomaterials. Graphene is atomically thin, chemically inert, consists of light atoms, and possesses a highly ordered structure. Graphene is electrically and thermally conductive, and is the strongest material ever measured.
We offer ultrahigh-quality graphene that is produced through the substrate-free gas-phase method which results in a highly ordered structure that is composed of 99% carbon by mass (1% hydrogen).
Mesh: 200
Material: Nickel
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.