Learn More
Cytiva Grade 1450CV Application-Specific Filters
For identification of undissolved dyes in the textile industry
Supplier: Cytiva 10313209
Description
- Basis Weight: 120
- Filtration Speed (Herzberg): 30
Specifications
Cellulose | |
1450 CV | |
Application-Specific Filter |
Identification of Undissolved Dyes in The Textile Industry | |
50/Pk. | |
90 mm |