Learn More
Electron Microscopy Sciences Goniometer Bases, Type B1
Supplier:  Electron Microscopy Sciences 6014001B1
Encompass_Preferred
Goniometer Bases tightly capture the MicroRT tubing for seamless room and low-temperature data collection.
Catalog No. 50-189-8869