Learn More
Perkin Elmer US LLC Gold Sample Pans and Covers - Qty. 10
Supplier: Perkin Elmer US LLC 02190042
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The Gold Sample Pans and Covers can be used for non-metal samples such as ceramics, coal, soils, or minerals. They can also be used for any materials that reacts with aluminum or when a sample must be heated over 600 °C.