Learn More
TARGETMOL CHEMICALS INC GLYH-101 10MG
Supplier: TARGETMOL CHEMICALS INC T2451
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GlyH-101 is a cell-permeable glycinyl hydrazone compound that blocks CFTR with Ki of 1.4 uM. purity: 98%