Learn More
TARGETMOL CHEMICALS INC GIREDESTRANT TARTRATE 5MG
Supplier: TARGETMOL CHEMICALS INC T11236
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Giredestrant tartrate (Estrogen receptor antagonist 1) is a novel orally active selective and effective non-steroidal estrogen receptor antagonist. Giredestrant tartrate strongly binds to ER resulting in the failure of ER to activate the transcription of targeted genes and promoting the degradation of ER protein. Giredestrant tartrate can be used to treat tumor diseases. purity: 98%
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.