Learn More
Electron Microscopy Sciences Variable Size Geological Thin Section Holder, 9.5mm Pin
Supplier: Electron Microscopy Sciences 7594116
This thin section holder is made from vacuum grade aluminum with copper clips and brass screws . The unit measures 51x32x8 mm and holds either standard thin sections of 47 x 27 mm or smaller or larger thin sections. Smallest size would be 40x20 mm. The holder comes complete with 2 clips to insure nothing moves. The holder has convenient tweezer inserts which allow for easy loading.
Type: 9.5mm Pin
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.