Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agilent BioTek Gen5 Microplate Reader and Imager Software

For use with Imaging Multi-Mode Readers.

Supplier:  Agilent BioTek GENSECPRIME

Request Bulk Order

 View more versions of this product

Catalog No. BTGENSCPRIM

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Explore available promotions
This item is not returnable. View return policy

Description

Description

  • Offers a unique combination of power and ease-of-use that drives productivity and saves time
  • Helps transform raw data and images into meaningful results
  • Performs high magnification, intracellular image analysis to basic ELISA
Gather and Analyze Data Quickly and Easily
  • Quick start with >150 prepared protocols available in Gen5 and online
  • Endpoint, kinetic, well area scanning, spectral scanning read modes
  • One-button quick export sends data directly to Excel, no need for multiple steps
  • Predefined Take3 Micro-Volume Plate protocols for direct quantification of 2μL samples
Explore, Image and Analyze Intra-and Inter-cellular Activity
  • Acquire high quality images from 1.25x to 100x with Lionheart FX and up to 60x with Cytation
  • Combine image acquisition steps and multi-mode detection steps to rapidly obtain and analyze phenotypic and quantitative data
  • Advanced features including image stitching, z-projection and digital phase contrast
  • Import image files from other programs for analysis in Gen5
Imaging: Methods and Processes
  • Automated cell counting and analysis with image statistics
  • Time-lapse movie making with automated image alignment
  • Montage capture, image stitching
  • Z-stacking and z-projection
  • High speed imaging, image binning for fast kinetics
QC and Validation Tools
  • Fully validated at Agilent BioTek
  • Gen5 Validation Package provides an efficient step-by-step approach to facilitate internal validation
  • Compatible with the comprehensive microplate reader IQ/OQ/PQ (Product Qualification Packages) to help speed instrument qualification
  • Gen5 IVD and Gen5 IVD Image+ have QC trending capability, security features and validation protocols for convenience and efficiency
Gen5 Image Prime for Cell Imaging
  • Offers powerful image acquisition and analysis tool for use with Lionheart FX Automated Cell Imager and Cytation Cell Imaging Multi­Mode Readers
  • Cell counting, subpopulation analysis, advanced analysis features
  • Multi-mode and imaging steps combined in single protocol allow hit-picking for imaging efficiency, no need to collect unnecessary data
  • Image processing and analysis including Z-projection, image stitching, hitpicking and digital phase contrast, plus advanced features including secondary mask, dynamic thresholding and subpopulation analysis
  • Built-in movie maker for acquired kinetic imaging sequences
  • Fluorescence, brightfield, color brightfield and phase contrast image acquisition
  • Import image files from other programs for analysis in Gen5
GxP Compliance with Gen5 Secure
  • For regulated laboratory environments, Gen5 Secure offers functionality to help comply with the requirements of 21 CFR Part 11
  • Offers imaging control for Lionheart FX Automated Live Cell Imager and Cytation Cell Imaging Multi-Mode Readers
  • Includes image processing and analysis including z-projection, image stitching, hitpicking and digital phase contrast
  • Advanced features include secondary mask, dynamic thresholding and subpopulation analysis
  • Complete reader control and data analysis for all Agilent BioTek microplate readers
  • Includes features designed to help laboratories maintain critical GxP requirements including 21 CFR Part 11
  • Electronic signature for data verification, approval, authorization and other customizable events
  • Data, protocol and system audit trails track and record actions with customizable comments
  • Multi-level user permission assignment with highly granular list of protectable functions
  • Multiple groups, with multiple users and permissions are definable
  • User lockout and reset, password aging are all customizable
  • Email notifications for administrator-defined events
  • Single sign-on setup available
  • Multiple administrators can be assigned
Computer System Requirements
  • 64-bit version of Windows 7 or Windows 10 Professional
  • Intel Core i5 (4th generation) Processor or higher
  • Intel 8 USB Chipset or higher
  • 8 GB RAM or greater
  • 128 GB Hard Drive space or greater
  • Monitor resolution of 1600 x 900
  • Keyboard and mouse
Software (Lionheart FX requires Gen5 3.0 or higher)
  • Microsoft Internet Explorer v 9.0 or later (for online Help)
  • Microsoft Excel for Quick Export and PowerExport: Gen5 3.0: Excel 2007 (32-bit edition only) or Excel 2010 - 2016 (32- and 64-bit editions); Gen5 2.0: Excel 2007 - 2016 (32-bit editions only)
Connectivity
  • USB 3 port for camera
  • USB 2 or 3 port for instrument
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Imaging Multi-Mode Readers
Gen5 Secure Image Prime
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.